(S)-Methyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-Lys(Z)-OMe.HCl
Formula:C₁₅H₂₃ClN₂O₄
MW:330.809 g/mol
MDL Number:MFCD00034846
CAS Number:27894-50-4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 330.809 g/mol
Storage Temperature:Refrigerator
EINECS:248-715-9
Customers Who Bought This Also Bought