(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: H-Lys(Z)-OBzl.HCl,H-Lys(Z)-OBzl·HCl
Formula:C₂₁H₂₆N₂O₄·HCl
MW:406.9 g/mol
MDL Number:MFCD00038981
CAS Number:6366-70-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 406.9 g/mol
:(S)-Benzyl-2-amino-6-(((benzyloxy)carbonyl)amino)hexanoate hydrochloride
Storage Temperature:Refrigerator
EINECS:228-859-9
Customers Who Bought This Also Bought