O-tert-Butyl-L-threonine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: (2S,3R)-2-Amino-3-(tert-butoxy)butanoic acid
Formula:C₈H₁₇NO₃
MW:175.23 g/mol
MDL Number:MFCD00037766
CAS Number:4378-13-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 175.23 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought