N-Benzoyl-L-tyrosine ethyl ester
Catalog #: E-1530.0025BA
Supplier: Bachem Americas
CAS Number: 3483-82-7
Specifications
- Size:25 g
- Cat. no.:E-1530.0025BA
- Supplier No.:E-1530.0025
Specifications
About this item
Synonyms: BTEE
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 313.35 g/mol
Formula:C₁₈H₁₉NO₄
MW:313.35 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00002388
CAS Number:3483-82-7
EINECS:222-469-2
Customers Who Bought This Also Bought