Nɑ-Acetyl-L-lysine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Nalpha-Acetyl-L-lysine
Formula:C₈H₁₆N₂O₃
MW:188.23 g/mol
MDL Number:MFCD00008233
CAS Number:1946-82-3
:Nɑ-Acetyl-L-lysine
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 188.23 g/mol
Storage Temperature:Ambient
EINECS:217-747-5
Customers Who Bought This Also Bought