Fmoc-Leu 0.5-0.8 mmol/g on Wang resin 100-200 mesh
Catalog #: D-2350.0005BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:5 g
- Cat. no.:D-2350.0005BA
- Supplier No.:D-2350.0005
Specifications
About this item
:Fmoc-Leu
:Length:
:Width:
:Height:
:Ø
:Thickness:
:100-200 mesh
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Storage Temperature:Ambient
MDL Number:MFCD00801260
Customers Who Bought This Also Bought