Fmoc-Asp(OtBu) 0.50-0.80 mmol/g on Wang resin 100-200 mesh
Supplier: Bachem Americas
Certificates
About this item
MDL Number:MFCD00801229
:Length:
:Width:
:Height:
:Ø
:Thickness:
:100-200 mesh
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:Fmoc-Asp(OtBu)
Storage Temperature:Ambient
Customers Who Bought This Also Bought