2-Chlorotrityl chloride, polymer supported 200-400 mesh
Supplier: Bachem Americas
Certificates
About this item
Synonyms: 2-CTC resin
MDL Number:MFCD00161760
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
Storage Temperature:Freezer
Customers Who Bought This Also Bought