Fmoc-Cys(Trt) 0.40-1.00 mmol/g on Wang resin 200-400 mesh
Supplier: Bachem Americas
Certificates
About this item
MDL Number:MFCD00801374
:Length:
:Width:
:Height:
:Ø
:Thickness:
:200-400 mesh
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:Fmoc-Cys(Trt)
Storage Temperature:Ambient
Customers Who Bought This Also Bought