Fmoc-Asp(OtBu) 0.50-0.80 mmol/g on Wang resin 200-400 mesh
Catalog #: D-1055.0001BA
Supplier: Bachem Americas
CAS Number:
Specifications
- Size:1 g
- Cat. no.:D-1055.0001BA
- Supplier No.:D-1055.0001
Specifications
About this item
:Length:
:Width:
:Height:
:Ø
:Thickness:
:200-400 mesh
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:Fmoc-Asp(OtBu)
Storage Temperature:Refrigerator
MDL Number:MFCD00801229
Customers Who Bought This Also Bought