Z-Asu(Otbu)-OH·DCHA
Catalog #: C-4155.0001BA
Supplier: Bachem Americas
CAS Number: 49645-27-4
Specifications
- Size:1 g
- Cat. no.:C-4155.0001BA
- Supplier No.:C-4155.0001
Specifications
About this item
Synonyms: Z-Asu(OtBu)-OH·DCHA
:Z-Asu(Otbu)-OH·DCHA
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 560.776 g/mol
Formula:C₃₂H₅₂N₂O₆
MW:560.776 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00038886
CAS Number:49645-27-4
Customers Who Bought This Also Bought