Z-N-(N-β-boc-aminoethyl)-Gly-OH
Catalog #: C-3865.0005BA
Supplier: Bachem Americas
CAS Number: 34046-07-6
Specifications
- Size:5 g
- Cat. no.:C-3865.0005BA
- Supplier No.:C-3865.0005
Specifications
About this item
Synonyms: Z-N-(N-β-Boc-aminoethyl)-Gly-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 352.387 g/mol
Formula:C₁₇H₂₄N₂O₆
MW:352.387 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00672345
CAS Number:34046-07-6
Customers Who Bought This Also Bought