Z-ARG(Boc)2-OH CHA
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Z-Arg(Boc)₂-OH·CHA
Formula:C₃₀H₄₉N₅O₈
MW:607.749 g/mol
MDL Number:MFCD00237325
CAS Number:145315-39-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 607.749 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought