Z-Dab(Boc)-OH·DCHA
Catalog #: C-3510.0005BA
Supplier: Bachem Americas
CAS Number: 3350-13-8
Specifications
- Size:5 g
- Cat. no.:C-3510.0005BA
- Supplier No.:C-3510.0005
Specifications
About this item
Synonyms: (S)-2-Benzyloxycarbonylamino-4-(Boc-amino)butyric acid dicyclohexylammonium salt
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 533.71 g/mol
Formula:C₂₉H₄₇N₃O₆
MW:533.71 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00237341
CAS Number:3350-13-8
Customers Who Bought This Also Bought