Cgp 29287
Catalog #: C-3195.0005BA
Supplier: Bachem Americas
CAS Number: 93287-54-8
Specifications
- Size:5 mg
- Cat. no.:C-3195.0005BA
- Supplier No.:C-3195.0005
Specifications
About this item
Synonyms: Z-Arg-Arg-Pro-Phe-His-Sta-Ile-His-Lys(Boc)-OMe
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 1495.797 g/mol
Formula:C₇₂H₁₁₀N₂₀O₁₅
MW:1495.797 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00133577
CAS Number:93287-54-8
Customers Who Bought This Also Bought