Z-D-Arg(Z)₂-OH
Supplier: Bachem Americas
Certificates
About this item
Formula:C₃₀H₃₂N₄O₈
MDL Number:MFCD00237330
CAS Number:104321-57-5
:Z-D-Arg(Z)₂-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 576,61 g/mol
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought