N-Carbobenzoxy-L-proline
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Z-Pro-OH,Z-L-Prolin,N-Cbz-L-proline,N-Carbobenzyloxy-L-proline
Formula:C₁₃H₁₅NO₄
MW:249.27 g/mol
MDL Number:MFCD00003170
CAS Number:1148-11-4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 249.27 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought