Z-Arg(Mtr)-OH CHA
Catalog #: C-2250.0025BA
Supplier: Bachem Americas
CAS Number: 80745-09-1
Specifications
- Size:25 g
- Cat. no.:C-2250.0025BA
- Supplier No.:C-2250.0025
Specifications
About this item
Synonyms: Z-Arg(Mtr)-OH.CHA
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 619.778 g/mol
:Z-Arg(Mtr)-OH CHA
Formula:C₂₄H₃₂N₄O₇S·C₆H₁₃N
MW:619.778 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00057997
CAS Number:80745-09-1
Customers Who Bought This Also Bought