Z-Hyp-OH
Catalog #: C-2015.0025BA
Supplier: Bachem Americas
CAS Number: 13504-85-3
Specifications
- Size:25 g
- Cat. no.:C-2015.0025BA
- Supplier No.:C-2015.0025
Specifications
About this item
Synonyms: N-Benzyloxycarbonyl-4-hydroxy-L-proline,Z-L-4-Hydroxyproline,trans-N-Cbz-4-hydroxy-L-proline,(2S,4R)-1-((Benzyloxy)carbonyl)-4-hydroxypyrrolidine-2-carboxylic acid
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 265.3 g/mol
:Z-Hyp-OH
Formula:C₁₃H₁₅NO₅
MW:265.3 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00037329
CAS Number:13504-85-3
EINECS:236-831-2
Customers Who Bought This Also Bought