N-Carbobenzoxy-L-aspartic acid
Catalog #: C-1325.0100BA
Supplier: Bachem Americas
CAS Number: 1152-61-0
Specifications
- Size:100 g
- Cat. no.:C-1325.0100BA
- Supplier No.:C-1325.0100
Specifications
About this item
Synonyms: N-Cbz-L-aspartic Acid,N-Carbobenzyloxy-L-aspartic acid
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 267.24 g/mol
Formula:C₁₂H₁₃NO₆
MW:267.24 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00002719
CAS Number:1152-61-0
EINECS:214-568-4
Customers Who Bought This Also Bought