(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)butanoic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Dab(ivDde)-OH
Formula:C₃₂H₃₈N₂O₆
MW:546.664 g/mol
MDL Number:MFCD01631660
CAS Number:607366-21-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 546.664 g/mol
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought