Fmoc-Lys(Dabsyl)-OH
Catalog #: B-3870.0001BA
Supplier: Bachem Americas
CAS Number: 185503-97-3
Specifications
- Size:1 g
- Cat. no.:B-3870.0001BA
- Supplier No.:B-3870.0001
Specifications
About this item
:Fmoc-Lys(Dabsyl)-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 655,77 g/mol
Formula:C₃₅H₃₇N₅O₆S
Storage Temperature:Freezer
MDL Number:MFCD01632021
CAS Number:185503-97-3
Customers Who Bought This Also Bought