Fmoc-Lys(ME,Boc)-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Lys(Boc)(Me)-OH,Fmoc-Lys(Me
Formula:C₂₇H₃₄N₂O₆
MW:482.577 g/mol
MDL Number:MFCD01861332
CAS Number:951695-85-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 482.577 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought