Fmoc-O-(benzylphospho)-L-serine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Ser(PO(OBzl)OH)-OH
Formula:C₂₅H₂₄NO₈P
MW:497.44 g/mol
MDL Number:MFCD00797869
CAS Number:158171-14-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 497.44 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought