(S)-2-(Fmoc-amino)-3-(2,2-dimethyl-4H-benzo[1,3]dioxin-6-yl)propionic acid
Catalog #: B-3310.0001BA
Supplier: Bachem Americas
CAS Number: 252049-13-1
Specifications
- Size:1 g
- Cat. no.:B-3310.0001BA
- Supplier No.:B-3310.0001
Specifications
About this item
Synonyms: Fmoc-β-(2,2-dimethyl-4H-benzo[1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 473.525 g/mol
Formula:C₂₈H₂₇NO₆
MW:473.525 g/mol
Storage Temperature:Refrigerator
MDL Number:MFCD02259491
CAS Number:252049-13-1
Customers Who Bought This Also Bought