Fmoc-Lys(Me)₂-OH·HCl
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-α-Fmoc-N-ε-dimethyl-N-L-lysine hydrochloride
Formula:C₂₃H₂₉ClN₂O₄
MW:432.945 g/mol
MDL Number:MFCD05662353
CAS Number:252049-10-8
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 432.945 g/mol
:Fmoc-Lys(Me)₂-OH·HCl
Storage Temperature:Freezer
Customers Who Bought This Also Bought