Fmoc-Asp(ODmab)-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Asp(ODMAB)-OH
Formula:C₃₉H₄₂N₂O₈
MW:666.771 g/mol
MDL Number:MFCD00797875
CAS Number:269066-08-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 666.771 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought