Fmoc-β-(2-quinolyl)-Ala-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-β-(2-quinolyl)-Ala-OH
Formula:C₂₇H₂₂N₂O₄
MW:438.483 g/mol
MDL Number:MFCD01318745
CAS Number:214852-56-9
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 438.483 g/mol
:Fmoc-β-(2-quinolyl)-Ala-OH
Storage Temperature:Freezer
Customers Who Bought This Also Bought