(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-((4-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)-3-methylbutyl)amino)benzyl)oxy)-5-oxopentanoic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Glu(ODmab)-OH
Formula:C₄₀H₄₄N₂O₈
MW:680.7860 g/mol
MDL Number:MFCD00797873
CAS Number:268730-86-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 680.7860 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought