Fmoc-D-Arg(Boc)2-OH
Catalog #: B-2940.0001BA
Supplier: Bachem Americas
CAS Number: 214852-34-3
Specifications
- Size:1 g
- Cat. no.:B-2940.0001BA
- Supplier No.:B-2940.0001
Specifications
About this item
Synonyms: Fmoc-D-Arg(Boc)₂-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 596.681 g/mol
:Fmoc-D-Arg(Boc)2-OH
Formula:C₃₁H₄₀N₄O₈
MW:596.681 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00798643
CAS Number:214852-34-3
Customers Who Bought This Also Bought