Fmoc-Lys(Dnp)-OH
Catalog #: B-2885.1000BA
Supplier: Bachem Americas
CAS Number: 148083-64-1
Specifications
- Size:1 g
- Cat. no.:B-2885.1000BA
- Supplier No.:B-2885.1000
Specifications
About this item
Synonyms: Nepsilon-2
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 534,525 g/mol
Formula:C₂₇H₂₆N₄O₈
Storage Temperature:Freezer
MDL Number:MFCD00273454
CAS Number:148083-64-1
Customers Who Bought This Also Bought