Fmoc-Lys(biotinyl-ε-aminocaproyl)-OH
Catalog #: B-2580.1000BA
Supplier: Bachem Americas
CAS Number: 160158-05-4
Specifications
- Size:1 g
- Cat. no.:B-2580.1000BA
- Supplier No.:B-2580.1000
Specifications
About this item
Synonyms: Fmoc-Lys(biotinyl-epsilon-aminocaproyl)-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 707.89 g/mol
:Fmoc-Lys(biotinyl-ε-aminocaproyl)-OH
Formula:C₃₇H₄₉N₅O₇S
MW:707.89 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00672715
CAS Number:160158-05-4
Customers Who Bought This Also Bought