Fmoc-asp-ofm
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Asp-Ofm
Formula:C₃₃H₂₇NO₆
MW:533.58 g/mol
MDL Number:MFCD00237653
CAS Number:187671-16-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 533.58 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought