N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-Fmoc-L-aspartic Acid
Formula:C₁₉H₁₇NO₆
MW:355.35 g/mol
MDL Number:MFCD00237654
CAS Number:119062-05-4
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 355.35 g/mol
:N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid
Storage Temperature:Ambient
Customers Who Bought This Also Bought