Fmoc-Lys(ipr,Boc)-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Nepsilon-Boc-Nalpha-Fmoc-Nepsilon-isopropyl-L-lysine,N-Fmoc-N'-boc-N'-isopropyl-L-Lysine
Formula:C₂₉H₃₈N₂O₆
MW:510.631 g/mol
MDL Number:MFCD00080276
CAS Number:201003-48-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 510.631 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought