Fmoc-Cys(SO₃H)-OH·disodium salt
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Cys(SO3H)-OH·disodium salt
Formula:C₁₈H₁₅NNa₂O₇S₂
MW:467.418 g/mol
MDL Number:MFCD00237016
CAS Number:163558-30-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 467.418 g/mol
:Fmoc-Cys(SO₃H)-OH·disodium salt
Storage Temperature:Freezer
Customers Who Bought This Also Bought