Fmoc-Dap(Fmoc)-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Nα-Fmoc-3-(Fmoc-amino)-L-alanine,(S)-2
Formula:C₃₃H₂₈N₂O₆
MW:548.595 g/mol
MDL Number:MFCD00237005
CAS Number:201473-90-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 548.595 g/mol
:Fmoc-Dap(Fmoc)-OH
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought