N-Fmoc-N'-trityl-D-glutamine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Nα-Fmoc-Nδ-trityl-D-glutamine
Formula:C₃₉H₃₄N₂O₅
MW:610.71 g/mol
MDL Number:MFCD00151924
CAS Number:200623-62-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 610.71 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought