Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-Lys-OH.HCl,Nɑ-Fmoc-L-lysine Hydrochloride
Formula:C₂₁H₂₄N₂O₄
MW:404.89 g/mol
MDL Number:MFCD00190889
CAS Number:139262-23-0
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 404.89 g/mol
:Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride
Storage Temperature:Refrigerator
Customers Who Bought This Also Bought