Fmoc-Lys(FOR)-OH
Catalog #: B-1865.0005BA
Supplier: Bachem Americas
CAS Number: 201004-23-1
Specifications
- Size:5 g
- Cat. no.:B-1865.0005BA
- Supplier No.:B-1865.0005
Specifications
About this item
:Fmoc-Lys(FOR)-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 396,443 g/mol
Formula:C₂₂H₂₄N₂O₅
Storage Temperature:Refrigerator
MDL Number:MFCD00237025
CAS Number:201004-23-1
Customers Who Bought This Also Bought