(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-Fmoc-L-aspartic Acid 1-tert-Butyl Ester,N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 1-tert-Butyl Ester,Fmoc-Asp(OH)-OtBu,Fmoc-Asp-OtBu
Formula:C₂₃H₂₅NO₆
MW:411.5 g/mol
MDL Number:MFCD00065631
CAS Number:129460-09-9
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 411.5 g/mol
:(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
Storage Temperature:Freezer
Customers Who Bought This Also Bought