Fmoc-Lys(Fmoc)-opfp
Catalog #: B-1675.0001BA
Supplier: Bachem Americas
CAS Number: 132990-14-8
Specifications
- Size:1 g
- Cat. no.:B-1675.0001BA
- Supplier No.:B-1675.0001
Specifications
About this item
Synonyms: Fmoc-Lys(Fmoc)-Opfp
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 756.726 g/mol
Formula:C₄₂H₃₃F₅N₂O₆
MW:756.726 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00153377
CAS Number:132990-14-8
Customers Who Bought This Also Bought