Fmoc-Val-OPfp
Catalog #: B-1565.0005BA
Supplier: Bachem Americas
CAS Number: 86060-87-9
Specifications
- Size:5 g
- Cat. no.:B-1565.0005BA
- Supplier No.:B-1565.0005
Specifications
About this item
Synonyms: N-Fmoc-L-valine pentafluorophenyl ester
:Fmoc-Val-OPfp
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 505,441 g/mol
Formula:C₂₆H₂₀F₅NO₄
MW:505.441 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00065687
CAS Number:86060-87-9
Customers Who Bought This Also Bought