N(α)-Fmoc-D-tryptophan
Catalog #: B-1450.0005BA
Supplier: Bachem Americas
CAS Number: 86123-11-7
Specifications
- Size:5 g
- Cat. no.:B-1450.0005BA
- Supplier No.:B-1450.0005
Specifications
About this item
Synonyms: Fmoc-D-Trp-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 426.46 g/mol
Formula:C₂₆H₂₂N₂O₄
MW:426.46 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00062954
CAS Number:86123-11-7
Customers Who Bought This Also Bought