N-Fmoc-D-leucine
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Fmoc-D-Leu-OH
Formula:C₂₁H₂₃NO₄
MW:353.4 g/mol
MDL Number:MFCD00062957
CAS Number:114360-54-2
:N-Fmoc-D-leucine
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 353.4 g/mol
Storage Temperature:Ambient
Customers Who Bought This Also Bought