Fmoc-Asp(OtBu)-OPfp
Catalog #: B-1110.0005BA
Supplier: Bachem Americas
CAS Number: 86061-01-0
Specifications
- Size:5 g
- Cat. no.:B-1110.0005BA
- Supplier No.:B-1110.0005
Specifications
About this item
Synonyms: N-Fmoc-L-aspartic acid 4-tert-butyl ester pentafluorophenyl ester
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 577,504 g/mol
:Fmoc-Asp(OtBu)-OPfp
Formula:C₂₉H₂₄F₅NO₆
MW:577.504 g/mol
Storage Temperature:Freezer
MDL Number:MFCD00037633
CAS Number:86061-01-0
Customers Who Bought This Also Bought