(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid
Supplier: Bachem Americas
Certificates
About this item
Synonyms: N-Fmoc-L-aspartic Acid 4-tert-Butyl Ester,4-tert-Butyl N-Fmoc-L-aspartate,Fmoc-Asp(OtBu)-OH
Formula:C₂₃H₂₅NO₆
MW:411.5 g/mol
MDL Number:MFCD00037131
CAS Number:71989-14-5
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 411.5 g/mol
Storage Temperature:Refrigerator
EINECS:276-251-7
Customers Who Bought This Also Bought