Boc-thionoSer(Bzl)-1-(6-nitro)benzotriazolide
Supplier: Bachem Americas
Certificates
About this item
Formula:C₂₁H₂₃N₅O₅S
MDL Number:MFCD01318727
CAS Number:184951-89-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 457,505 g/mol
Storage Temperature:Freezer
Customers Who Bought This Also Bought