Boc-D-Arg(Pmc)-OH
Catalog #: A-4185.0005BA
Supplier: Bachem Americas
CAS Number: 214630-02-1
Specifications
- Size:5 g
- Cat. no.:A-4185.0005BA
- Supplier No.:A-4185.0005
Specifications
About this item
:Boc-D-Arg(Pmc)-OH
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 540,676 g/mol
Formula:C₂₅H₄₀N₄O₇S
Storage Temperature:Refrigerator
MDL Number:MFCD00798617
CAS Number:214630-02-1
Customers Who Bought This Also Bought