Boc-Lys(ME)2-OH
Supplier: Bachem Americas
Certificates
About this item
Synonyms: Boc-Lys(Me)₂-OH
Formula:C₁₃H₂₆N₂O₄
MW:274.361 g/mol
MDL Number:MFCD00076965
CAS Number:65671-53-6
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 274.361 g/mol
:Boc-Lys(ME)2-OH
Storage Temperature:Freezer
Customers Who Bought This Also Bought